1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Catalog # EHERC10020000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:372.93 g/mol
Boiling Pt:167 °C (1013 hPa)
Melting Pt:78 °C
Storage Temperature:Ambient
EINECS:200-317-6
CAS Number:6001-64-5
Formula:C₈H₁₆Cl₆O₃
MDL Number:MFCD02179352