Corn oil
Catalog # EVOK7284.0250
Supplier: EVONIK MS PROJECT
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
Melting Pt:–10 °C
Density:0.918 g/cm³ (20 °C)
EINECS:232-281-2
CAS Number:8001-30-7
MDL Number:MFCD00130865