Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pepsin
Pepsin
Pepsin
Catalog # CSLO2501-250G
Supplier:
CAESAR LORENTZ
CAS Number:
Expand image
Pepsin
Catalog # CSLO2501-250G
Supplier:
CAESAR LORENTZ
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
250 g
CAS No.:
9001-75-6
Storage conditions:
Refrigerator
Enzyme name:
Pepsin
Enzyme activity:
70 FIP-U/mg
Specifications
About this item
Product Details
Product Details