Potassium sorbate
Glass bottle for solids, 25 g
Supplier: CAESAR LORENTZ
Jump to:
Product Details & Documents
Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
- CAS Number:24634-61-5
- Formula:CH₃CH=CHCH=CHCOOK
- MDL Number:MFCD00016546
- Molecular Weight:150.22 g/mol
- Merck Index:13,07756
- Melting Pt:270 °C
- Density:1.36 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:246-376-1
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



