Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol

3-Fluoropyrocatechol

Marketsource
Supplier:   Thermo Scientific
3-Fluoropyrocatechol
Supplier:  Thermo Scientific
Marketsource
Certificates
3-Fluoropyrocatechol

3 Options Available Below

Product Details & Documents
Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:71 °C
  • Density:1,514 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource