Sodium cholate, Millipore®
Catalog # 229101-250
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:430.56 g/mol
Melting Pt:198 °C
Storage Temperature:Ambient
EINECS:206-643-5
CAS Number:361-09-1
:206986-87-0
Formula:C₂₄H₃₉NaO₅
MDL Number:MFCD00150749