Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Tricine

3 Options Available Below

Product Details & Documents
Synonyms:  2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
  • CAS Number:5704-04-1
  • Formula:(HOCH₂)₃CNHCH₂CO₂H
  • MDL Number:MFCD00004277
  • Molecular Weight:179.17 g/mol
  • Merck Index:13,09724
  • Melting Pt:187 °C
  • Density:1.523 g/cm³ (19.85 °C)
  • Storage Temperature:Ambient
  • EINECS:227-193-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg