5-Methylphenazinium methyl sulphate
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:299-11-6
:
Formula:C₁₄H₁₄N₂O₄S
MDL Number:MFCD00011923
:306.34 g/mol
Merck Index:12,06185
Melting Pt:198 °C
EINECS:206-072-1
:GHS08
:GHS07
Storage Temperature:Freezer