Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole
Supplier:
BIONET RESEARCH
Certificates
Expand image
3-Bromo-7-nitro-1H-indazole
Supplier:
BIONET RESEARCH
About this item
CAS Number:
74209-34-0
:
Formula:
C₇H₄BrN₃O₂
MDL Number:
MFCD00159910
:
242.03 g/mol
Melting Pt:
176 °C