Acetic acid glacial for HPLC, J.T.Baker®
Catalog # 9515-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.7 %
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:= 0.05
- Assay (by GC, corrected for water):≥99.7 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.01
- Residue after Evaporation:≤0.0005 %
- Water (by Karl Fischer titrn):≤0.1 %
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152