Order Entry
Germany
ContactUsLinkComponent
Dodecyltrimethylammonium chloride 50% in 2-propanol, high purity
Flammable
Corrosive
Irritant
Dodecyltrimethylammonium chloride 50% in 2-propanol, high purity
Catalog # 0624-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Dodecyltrimethylammonium chloride 50% in 2-propanol, high purity
Catalog # 0624-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Activity ( conc.):
    49,0 - 52,0 %
  • Free Amine & Amine HCl:
    < 2,0 %
  • pH (10%, Water) at 25 °C:
    6,0 - 9,0
  • Purity (C₁₅H₃₄ClN):
    ≥98,5%
  • Specific Gravity:
    Report value

Specification Test Results

About this item

 :
 :263.89 g/mol
EINECS:203-927-0
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:112-00-5
Formula:C₁₅H₃₄ClN
MDL Number:MFCD00041974