Order Entry
Germany
Orders LinkContactUsLinkComponent
Ethoxytrimethylsilane ≥97%

Ethoxytrimethylsilane ≥97%

Glass bottle for solids, 100 g
Catalog # L10151.22
Flammable
Irritant
Ethoxytrimethylsilane ≥97%
Catalog # L10151.22
CAS Number:  1825-62-3
Ethoxytrimethylsilane ≥97%
Ethoxytrimethylsilane ≥97%

Glass bottle for solids, 100 g

CAS Number:  1825-62-3
Price
94,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 1825-62-3 Molecular Formula: C5H14OSi Formula Weight: 118.25 Flash Point: -18°(-0°F) Refractive Index: 1.3740 Odour: Mild Melting Point: -83° Boiling Point: 75-76° Beilstein Reference: 1731950 UN No.: 1993 MDL No.: MFCD00009069

Synonyms:  Trimethyl(ethoxy)silane
  • CAS Number:1825-62-3
  • Formula:C₅H₁₄OSi
  • MDL Number:MFCD00009069
  • Molecular Weight:118.25 g/mol
  • Boiling Pt:75…76 °C
  • Melting Pt:–83 °C
  • Density:0.757
  • Flash Pt:–18 °C (–0 °F)
  • Storage Temperature:Ambient
  • EINECS:217-370-6
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.