Phenyltrimethylsilane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:768-32-1
:
Formula:C₉H₁₄Si
MDL Number:MFCD00008271
Boiling Pt:169…170 °C
Density:0.871
Flash Pt:40 °C (104 °F)
EINECS:212-192-5
ADR:3,III
:150.3 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient