Order Entry
Germany
Orders LinkContactUsLinkComponent
 

9-Hydroxyfluorene ≥98%

25 g
Catalog # L01917.14
9-Hydroxyfluorene ≥98%
Catalog # L01917.14
CAS Number:  1689-64-1
9-Hydroxyfluorene ≥98%

25 g

CAS Number:  1689-64-1
Price
54,20 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 1689-64-1 Molecular Formula: C13H10O Formula Weight: 182.22 Melting Point: 155-157° Beilstein Reference: 1869799

Synonyms:  9-Fluorenol, Fluoren-9-ol
  • CAS Number:1689-64-1
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00001135
  • Molecular Weight:182.22 g/mol
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:155…157 °C
  • Density:1.151 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:216-879-0
Specifications

Specifications

  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.