

4-Bromobenzyl mercaptan ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19552-10-4
:
Formula:C₇H₇BrS
MDL Number:MFCD03701566
:203.1
Melting Pt:26…30 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
:GHS07