2-Bromthiazol ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 3034-53-5 Molecular Formula: C3H2BrNS Formula Weight: 164.02 Storage Temperature: 2°C to 8°C Flash Point: 63°(145°F) Refractive Index: 1.5930 Boiling Point: 171° Beilstein Reference: 105724 MDL No.: MFCD00005316
- CAS Number:3034-53-5
- Formula:C₃H₂BrNS
- MDL Number:MFCD00005316
- Molecular Weight:164.03 g/mol
- Boiling Pt:171 °C
- Density:1,836
- Flash Pt:63 °C (145 °F)
- EINECS:221-229-4
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 3034-53-5 Molecular Formula: C3H2BrNS Formula Weight: 164.02 Storage Temperature: 2°C to 8°C Flash Point: 63°(145°F) Refractive Index: 1.5930 Boiling Point: 171° Beilstein Reference: 105724 MDL No.: MFCD00005316
- CAS Number:3034-53-5
- Formula:C₃H₂BrNS
- MDL Number:MFCD00005316
- Molecular Weight:164.03 g/mol
- Boiling Pt:171 °C
- Density:1,836
- Flash Pt:63 °C (145 °F)
- EINECS:221-229-4
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



