0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Chloro-N-methylaniline
2-Chloro-N-methylaniline ≥97%
2-Chloro-N-methylaniline ≥97%
:
Thermo Fisher Scientific
2-Chloro-N-methylaniline ≥97%
:
Thermo Fisher Scientific
:
932-32-1
:
:
C₇H₈ClN
:
MFCD00045170
:
141.6 g/mol
:
218 °C
:
1.150
:
92 °C (197 °F)
:
2810
:
6.1,III
:
GHS06
:
GHS08
:
GHS07
:
Ambient
msds_ALFAA14343
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years