2-(4-Bromophenyl)ethanol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 4654-39-1 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 110°(230°F) Refractive Index: 1.5730 Boiling Point: 106-108°/1mm Beilstein Reference: 2079929 MDL No.: MFCD00002897
Synonyms: 4-Bromophenethyl alcohol
- CAS Number:4654-39-1
- Formula:C₈H₉BrO
- MDL Number:MFCD00002897
- Molecular Weight:201.06 g/mol
- Boiling Pt:106…108 °C (1 mmHg)
- Melting Pt:36…38 °C
- Density:1.436
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:225-093-7
4 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 4654-39-1 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 110°(230°F) Refractive Index: 1.5730 Boiling Point: 106-108°/1mm Beilstein Reference: 2079929 MDL No.: MFCD00002897
Synonyms: 4-Bromophenethyl alcohol
- CAS Number:4654-39-1
- Formula:C₈H₉BrO
- MDL Number:MFCD00002897
- Molecular Weight:201.06 g/mol
- Boiling Pt:106…108 °C (1 mmHg)
- Melting Pt:36…38 °C
- Density:1.436
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:225-093-7



