
Isopropyl-4-hydroxybenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4191-73-5
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00016468
:180.2 g/mol
Melting Pt:84…86 °C
EINECS:224-069-3
:GHS07
Storage Temperature:Ambient