
Iron(II) oxalate dihydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6047-25-2
:
Formula:Fe(C₂O₄)·2H₂O
MDL Number:MFCD00150040
:179.9 g/mol
Melting Pt:190 °C (dec.)
Density:2.28
Storage Temperature:Ambient
EINECS:208-217-4
UN:3288
ADR:6.1,III
:GHS07