Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethylisoxazole
3,5-Dimethylisoxazole ≥99%
3,5-Dimethylisoxazole ≥99%
:
Thermo Fisher Scientific
3,5-Dimethylisoxazole ≥99%
:
Thermo Fisher Scientific
:
300-87-8
:
:
C₅H₇NO
:
MFCD00003156
:
97.12 g/mol
:
142…144 °C
:
–14 °C
:
0.992
:
31 °C (87 °F)
:
Ambient
:
206-100-2
:
3,III
:
GHS02
msds_ALFAA12217
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years