

4-Fluorobenzyl mercaptan 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15894-04-9
:
Formula:C₇H₇FS
MDL Number:MFCD00040724
:142.2 g/mol
Boiling Pt:72…74 °C (15 mmHg)
Density:1.157
Flash Pt:76 °C (168 °F)
Storage Temperature:Ambient
EINECS:240-031-9
UN:2810
ADR:6.1,III
:GHS06
:GHS07