Ammonium hexachloroplatinate(IV) (≥43.4% Pt)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16919-58-7
:
Formula:H₈Cl₆N₂Pt
MDL Number:MFCD00010886
:443.87 g/mol
Merck Index:14,548
Melting Pt: Dec.
Density:3.06
EINECS:240-973-0
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient