

6-Chloro-1-hexyne 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10297-06-0
:
Formula:C₆H₉Cl
MDL Number:MFCD00013697
:116.59 g/mol
Flash Pt:37 °C
UN:1993
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4490 to 1.4510 (20°C, 589 nm)
- GC:≥96.0 %