
8-Bromocaprylic acid 95%
:17696-11-6
:
:BrH₂C(CH₂)₆COOH
:MFCD00004430
:223.11 g/mol
:147…150 °C (2 mmHg)
:36…41 °C
:1,324 g/cm³ (20 °C)
:>110 °C
:3261
:GHS05
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:36 to 41 °C
- GC:≥94 %