2-(2-Bromophenyl)ethanol 99%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:1074-16-4
- Formula:C₈H₉BrO
- MDL Number:MFCD00093566
- Molecular Weight:201.06 g/mol
- Boiling Pt:97 °C (.7 mmHg)
- Flash Pt:>110 °C
2 Options Available Below
Product Details & Documents
- CAS Number:1074-16-4
- Formula:C₈H₉BrO
- MDL Number:MFCD00093566
- Molecular Weight:201.06 g/mol
- Boiling Pt:97 °C (.7 mmHg)
- Flash Pt:>110 °C
Documents
ACRO35055
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5755 to 1.5775 (20°C, 589 nm)
- GC:≥98.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



