N-Propylethylenediamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-39-7
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008172
:102.18 g/mol
Boiling Pt:147…150 °C
Density:0.82 g/cm³
Flash Pt:14 °C
EINECS:203-864-9
UN:2733
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4394 to 1.4414 (20°C, 589 nm)
- GC:≥98.5 %