Ethoxytrimethylsilane 95%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Trimethyl(ethoxy)silane
- CAS Number:1825-62-3
- Formula:C₅H₁₄OSi
- MDL Number:MFCD00009069
- Molecular Weight:118.25 g/mol
- Boiling Pt:75…76 °C (760 mmHg)
- Melting Pt:–83 °C
- Density:0.75 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:217-370-6
- UN:1993
- ADR:3,II
3 Options Available Below
Product Details & Documents
Synonyms: Trimethyl(ethoxy)silane
- CAS Number:1825-62-3
- Formula:C₅H₁₄OSi
- MDL Number:MFCD00009069
- Molecular Weight:118.25 g/mol
- Boiling Pt:75…76 °C (760 mmHg)
- Melting Pt:–83 °C
- Density:0.75 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:217-370-6
- UN:1993
- ADR:3,II
Documents
ACRO31382
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3730 to 1.3750 (20°C, 589 nm)
- GC:≥94 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



