1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6001-64-5
:
Formula:C₈H₁₆Cl₆O₃
MDL Number:MFCD02179352
:372.93 g/mol
Boiling Pt:173…175 °C (760 mmHg)
Melting Pt:75…79 °C
Flash Pt:100 °C
Storage Temperature:Ambient
EINECS:200-317-6
:GHS07
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:97.5 to 102.5 % (ex Cl)
- Water:4.5 to 5.5 % (K.F.)