Chemical Drum Spill Kit
Catalog # BRDY813877
Supplier: Brady
Specifications
- Weight:47 kg
- Colour:Yellow
- Description:Chemical drum spill kit, SKH-95
- Absorbency:275 L
- Pk:1 KIT
Specifications
About this item
The drum spill kit is designed for large spills of chemicals and hazardous liquids.
- Drum spill kit that comes in a polyethylene, screw top container
- Water and chemical resistant to protect the sorbent material
- Drum can also be used as shipping container for used sorbents
- Moulded design allows for easy gripping and pick up by a forklift or trolley
Delivery: Supplied with 100 pads (410×510 mm), 8 socs (Ø 7,6×3660 mm), 12 socs (Ø 7,6×1220 mm), 8 pillows (430×480 mm), 50 wipes (300×380 mm), 1 pair of gloves, 1 pair of goggles and 10 disposal bags.