Ninhydrin solution in 2-propanol, spray reagent for TLC, Supelco®
Catalog # 1.06705.0100
Supplier: Merck
CAS Number:
Specifications
- Pk:100 ml
- Seq:2440
Specifications
About this item
:
:178.14 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:1219
:GHS02
:GHS07
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791