Orders LinkContactUsLinkComponent
Skull, fetal
Skull, fetal
Fetal skull
Catalog # 763-0077
Marketsource
Supplier:   3B Scientific
Skull, fetal
Catalog # 763-0077
Supplier:  3B Scientific
Marketsource
Skull, fetal
Skull, fetal
Fetal skull
Price
98,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
The anatomy of a human fetal skull can be studied easily with this realistic fetal skull replica.
The fetal skull model is a natural cast of a fetal head in the 30th week of pregnancy showing the characteristics of prenatal development.
The fontanelle, which becomes bone over time, are clearly visible on the fetal skull.
Sutures will form along the bony plates helping fuse the fetal skull as the child ages.
Specifications
  • Description:
    Fetal skull