Order Entry
Germany
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-L-(-)-proline

cis-4-Hydroxy-L-(-)-proline

Glass bottle for solids, 1 g
Catalog # MOLE28587393-1G
Marketsource
Supplier:   Molekula
cis-4-Hydroxy-L-(-)-proline
Catalog # MOLE28587393-1G
Supplier:  Molekula
Marketsource
CAS Number:  618-27-9
cis-4-Hydroxy-L-(-)-proline

Glass bottle for solids, 1 g

CAS Number:  618-27-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:257 °C
  • EINECS:210-542-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.