Order Entry
Germany
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-L-(-)-proline

cis-4-Hydroxy-L-(-)-proline

Marketsource
Supplier:   BACHEM BIOCHEMICA
cis-4-Hydroxy-L-(-)-proline
Supplier:  BACHEM BIOCHEMICA
Marketsource
Certificates
cis-4-Hydroxy-L-(-)-proline

2 Options Available Below

Product Details & Documents
Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:257 °C
  • EINECS:210-542-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource