Order Entry
Germany
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-L-(-)-proline ≥99%

cis-4-Hydroxy-L-(-)-proline ≥99%

cis-4-Hydroxy-L-(-)-proline ≥99%
Certificates
cis-4-Hydroxy-L-(-)-proline ≥99%
cis-4-Hydroxy-L-(-)-proline ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 618-27-9 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: ca 250° dec. Optical Rotation: -59° (c=2 in water)

Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:∼ 250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:210-542-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 222,00 €
    222,00 €
    1 * 250 mg
       222,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 639,00 €
    639,00 €
    1 * 1 g
       639,00 €