Order Entry
Germany
ContactUsLinkComponent
cis-4-Hydroxy-D(+)-proline ≥98%
cis-4-Hydroxy-D(+)-proline ≥98%
Catalog # H27633.03
CAS Number:  
cis-4-Hydroxy-D(+)-proline ≥98%
Catalog # H27633.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :131.13 g/mol
Melting Pt:243 °C (dec.)
Storage Temperature:Ambient
EINECS:219-963-5
CAS Number:2584-71-6
Formula:C₅H₉NO₃
MDL Number:MFCD00005252