Order Entry
Germany
ContactUsLinkComponent
(S)-(+)-Mandelic acid
(S)-(+)-Mandelic acid
Catalog # MOLE41911890-25G
Supplier:  Molekula
CAS Number:  
(S)-(+)-Mandelic acid
Catalog # MOLE41911890-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:322 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1,349 g/cm³ (20 °C)
Flash Pt:>190 °C
 :152.15 g/mol
EINECS:241-240-8
 :GHS05
CAS Number:17199-29-0
Formula:C₈H₈O₃
MDL Number:MFCD00004495