Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Serine Hydrazide Hydrochloride
DL-Serine Hydrazide Hydrochloride 97%
DL-Serine Hydrazide Hydrochloride 97%
DL-Serine Hydrazide Hydrochloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
DL-Serine Hydrazide Hydrochloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
55819-71-1
:
Formula:
C₃H₁₀ClN₃O₂
MDL Number:
MFCD08704793
:
155.58 g/mol
Melting Pt:
183 °C
:
GHS07