Order Entry
Germany
ContactUsLinkComponent
1,3-Oxazolo[4,5-b]pyridine-2-thiol
1,3-Oxazolo[4,5-b]pyridine-2-thiol
Catalog # APOSOR14288-100MG
Supplier:  Apollo Scientific
CAS Number:  
1,3-Oxazolo[4,5-b]pyridine-2-thiol
Catalog # APOSOR14288-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :152.174 g/mol
Melting Pt:241 - 243 °C
 :GHS07
CAS Number:211949-57-4
 :53052-06-5
Formula:C₆H₄N₂OS
MDL Number:MFCD00973914
Melting Pt:241…243 °C