

2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27776-21-2
:
Formula:C₁₂H₂₄Cl₂N₆
MDL Number:MFCD00142723
:323.27 g/mol
Melting Pt:190 °C
:GHS05
:GHS07
Storage Temperature:Refrigerator