Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodoacetamide
Iodoacetamide 98%
Iodoacetamide 98%
:
Apollo Scientific
Iodoacetamide 98%
:
Apollo Scientific
:
144-48-9
:
:
ICH₂CONH₂
:
MFCD00008028
:
184.96 g/mol
:
92…95 °C
:
2811
:
6.1,III
:
GHS06
:
GHS08
:
Refrigerator