meso-2,3-Dibromosuccinic acid, Sigma-Aldrich®
Catalog # SIAL105473-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:13,03054
Boiling Pt:352 °C (1013 hPa)
Melting Pt:272 °C
UN:3261
ADR:8,III
:M-Clarity™ quality level = MQ200
:275.88 g/mol
EINECS:452-290-7
CAS Number:608-36-6
Formula:HOOC(BrCH)₂COOH
MDL Number:MFCD00066439