
meso-2,3-Dibromosuccinic acid 98%
Catalog # ACRO147575000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance:White to yellow to light beige Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %
- Specific optical rotation:+54.0° to +58.0° (20°C, 589 nm) (c=1, acetone)
Specification Test Results
About this item
:
Merck Index:13,03054
Boiling Pt:352 °C (1013 hPa)
Melting Pt:>275 °C (subl.)
Storage Temperature:Ambient
ADR:8,III
:275.88 g/mol
EINECS:452-290-7
:GHS07
CAS Number:608-36-6
Formula:HOOC(BrCH)₂COOH
MDL Number:MFCD00066439