0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
9,10-Anthraquinone-2-carboxylic acid
9,10-Anthraquinone-2-carboxylic acid 95%
9,10-Anthraquinone-2-carboxylic acid 95%
APOSOR7750-5G
:
Apollo Scientific
:
9,10-Anthraquinone-2-carboxylic acid 95%
APOSOR7750-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
252.22 g/mol
:
287 - 289 °C
:
GHS07
:
117-78-2
:
C₁₅H₈O₄
:
MFCD00001231
:
287…289 °C