Sterile Goods Baskets for ISO Units
Supplier: HUPFER METALLWERKE
Certificates
About this item
Sterile supplies basket with insert frame for transporting, storing, and presenting sterile supplies in soft packaging without the use of containers.
- Produced from electro-polished stainless steel
- With U-shape profile rail compatible drawer frame
- Stackable inside and on top of each other
Basket suitable for washing and sterilising devices, made from premium electro-polished round stainless steel material. Basket with upper insert frame suitable for sliding into L or U profile rails of cabinet systems, Norm 5 Medidul shelves or Hupfer Känguruh System®. The special design of the faces means the baskets can be stacked either one inside the other (when empty, for example during washing and disinfecting), or on top of each other (when filled).