Orders LinkContactUsLinkComponent
Measuring Cables, CYK10
Measuring Cables, CYK10
Supplier:   Endress+Hauser
Measuring Cables, CYK10
Supplier:  Endress+Hauser
Certificates
Measuring Cables, CYK10
Measuring Cables, CYK10
3 Options Available Below
Product Details & Documents
CYK10 measuring cable is used with all sensors with Memosens plug-in head. Forget about moisture and benefit from completely safe signal transmission.
  • Memosens data cable for all sensors with Memosens plug-in head
  • Maximum process safety via non-contact, inductive and digital signal transmission
  • No contact corrosion, absolutely watertight, no interference potential
  • Increased availability of the measuring point: Digital data transmission provides an automatic error message if the signal is lost
CYK10 guarantees absolute water-proof connection of Memosens sensors to the transmitter. Its inductive coupling shows no contact corrosion and guarantees galvanic isolation of transmitter and medium. Simply forget about limited cable length, interfering potentials or moisture and benefit from completely safe and reliable signal transmission.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • ENDRCYK10-A031
    Measuring cable CYK10, wire terminals; Nonhazardous area approval
    Measuring cable CYK10, wire terminals; Nonhazardous area approval
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 192,00 €
    192,00 €
    1 * 1 ST
       192,00 €
  • ENDRCYK10-A051
    Measuring cable CYK10, wire terminals; Nonhazardous area approval
    Measuring cable CYK10, wire terminals; Nonhazardous area approval
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • ENDRCYK10-A101
    Measuring cable CYK10, wire terminals; Nonhazardous area approval
    Measuring cable CYK10, wire terminals; Nonhazardous area approval
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 288,00 €
    288,00 €
    1 * 1 ST
       288,00 €