(±)-Mandelic acid for synthesis, Sigma-Aldrich®
: Merck
:90-64-2
:C₈H₈O₃
:MFCD00064250
:152.15 g/mol
:322 °C (1013 hPa)
:117…120 °C
:1.361 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:210-277-1
:GHS05
- Solubility:150 g/l (20 °C)
- Melting Point:117 - 120 °C
- Molar Mass:152.15 g/mol
- Bulk Density:750 kg/m3
- Density:0.7 - 0.8 g/cm3 (20 °C)
- pH:2.3 (10 g/l, H2O)
- Synonyms:DL-α-Hydroxyphenylacetic acid, Phenylglycollic acid
- Hill Formula:C8H8O3
- Chemical Formula:C6H5CH(OH)COOH