(±)-4-Hydroxy-3-methoxymandelic acid
Catalog # PROOMM0262.06
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:198.18 g/mol
Merck Index:12,10070
Boiling Pt:421 °C (1013 hPa)
Melting Pt:132…134 °C
Density:1.44 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-224-0
:GHS07
CAS Number:55-10-7
:2394-20-9
Formula:C₉H₁₀O₅
MDL Number:MFCD00004235