Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lecithin from egg yolk
Lecithin from egg yolk ≥96%, powder
Lecithin from egg yolk ≥96%, powder
:
MP Biomedicals
Lecithin from egg yolk ≥96%, powder
:
MP Biomedicals
:
93685-90-6
:
:
MFCD00131449
:
Refrigerator
:
297-639-2