Order Entry
Germany
ContactUsLinkComponent
ICG alkyne fluorescent probe
Irritant
ICG alkyne fluorescent probe
Catalog # AATB986
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
ICG alkyne fluorescent probe
Catalog # AATB986
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :768.03 g/mol
ADR:5,II
 :GHS07
CAS Number:1622335-40-3